Search
Search

Fut8| Gene Symbol: Fut8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002289.1 | NP_001002289.1 | 9 | 1581 | ||
| XM_008764825.1 | XP_008763047.1 | 12 | 2089 | |||
| XM_008764826.1 | XP_008763048.1 | 12 | 3009 | |||
| GenBank mRNA | AJ781406.1 | - | 9 | 1581 | ||
| FN668936.1 | - | 12 | 2089 | |||
| FN668937.1 | - | 12 | 2118 | |||
| FN668938.1 | - | 9 | 1803 | |||