Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013627.2 | NP_001013649.2 | 6 | 2728 | ||
XM_011530932.1 | XP_011529234.1 | 7 | 2812 | |||
XM_011530933.1 | XP_011529235.1 | 6 | 2680 | |||
XM_011530934.2 | XP_011529236.1 | 6 | 7520 | |||
XM_017029473.1 | XP_016884962.1 | 6 | 6490 | |||
XM_017029474.1 | XP_016884963.1 | 6 | 2632 | |||
XM_017029475.1 | XP_016884964.1 | 2 | 5875 | |||
XM_017029476.1 | XP_016884965.1 | 2 | 1992 | |||
XM_017029477.1 | XP_016884966.1 | 2 | 6906 | |||
GenBank mRNA | BC033261.1 | - | 2 | 2892 | ||
BC136756.1 | - | 3 | 2101 | 9020746 |