Search
Search

Fggy| Gene Symbol: Fggy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113412.1 | NP_001106883.1 | 7, 8 | 898 | ||
| NM_029347.2 | NP_083623.1 | 7, 8 | 947 | |||
| GenBank mRNA | AK009249.1 | - | 1, 2 | 68 | ||
| AK085534.1 | - | 7, 8 | 909 | |||
| AK138439.1 | - | 7, 8 | 895 | |||
| BC031708.1 | - | 7, 8 | 865 | 4194716 | ||