Search
Search

PROX2| Gene Symbol: PROX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080408.2 | NP_001073877.2 | 3 | 2003 | ||
| NM_001243007.1 | NP_001229936.1 | 4 | 2684 | |||
| GenBank mRNA | BC105720.1 | - | 4 | 2683 | ||
| BC105927.1 | - | 4 | 2683 | |||