Search
Search

ADAMTS20| Gene Symbol: ADAMTS20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025003.3 | NP_079279.3 | 2 | 410 | ||
| XM_011538754.2 | XP_011537056.1 | 2 | 460 | |||
| GenBank mRNA | AF488804.1 | - | 2 | 410 | ||
| AJ515153.3 | - | 2 | 410 | |||
| AJ515154.2 | - | 2 | 410 | |||