Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000850.4 | NP_000841.1 | 6 | 709 | ||
NM_147148.2 | NP_671489.1 | 6 | 709 | |||
NR_024538.1 | 5 | 627 | ||||
XM_017001085.1 | XP_016856574.1 | 6 | 709 | |||
XM_017001086.1 | XP_016856575.1 | 6 | 941 | |||
XM_017001087.1 | XP_016856576.1 | 6 | 935 | |||
XM_017001088.1 | XP_016856577.1 | 5 | 620 | |||
GenBank mRNA | AK291880.1 | - | 6 | 709 | ||
BC015513.1 | - | 6 | 669 | 3893061 | ||
BC108729.1 | - | 6 | 666 | 6171894 | ||
BI458242.1 | - | 6 | 710 | |||
CR541869.1 | - | 6 | 395 | |||
DQ062813.1 | - | 4 | 497 | |||
M96234.1 | - | 6 | 658 | |||
M99421.1 | - | 5 | 451 | |||
M99422.1 | - | 6 | 667 |