Search
Search

ADGRE3| Gene Symbol: ADGRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032571.4 | NP_115960.2 | 4 | 424 | ||
| XM_011528374.2 | XP_011526676.1 | 3 | 315 | |||
| GenBank mRNA | AF239764.1 | - | 4 | 361 | ||
| AK290909.1 | - | 4 | 348 | |||
| AL832663.1 | - | 4 | 425 | |||
| BC121130.2 | - | 4 | 346 | |||
| BC121131.2 | - | 4 | 346 | |||
| BC128053.1 | - | 4 | 346 | |||
| BC148282.2 | - | 4 | 346 | |||
Set Membership: |
|