Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013168.2 | NP_037300.2 | 6 | 432 | ||
XM_006242896.1 | XP_006242958.1 | 6 | 518 | |||
XM_008766148.1 | XP_008764370.1 | 5 | 602 | |||
XM_008766149.1 | XP_008764371.1 | 5 | 539 | |||
GenBank mRNA | BC088162.1 | - | 6 | 432 | 7375228 | |
FQ226062.1 | - | 6 | 355 | |||
FQ231091.1 | - | 6 | 416 | |||
FQ234870.1 | - | 4 | 588 | |||
X06827.1 | - | 6 | 307 | |||
Y12006.1 | - | 6 | 302 |