Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nanog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289828.1 | NP_001276757.1 | 4 | 991 | ||
NM_001289830.1 | NP_001276759.1 | 5 | 791 | |||
NM_001289831.1 | NP_001276760.1 | 5 | 775 | |||
NM_028016.3 | NP_082292.1 | 4 | 994 | |||
GenBank mRNA | AB093574.1 | - | 4 | 965 | ||
AF507043.1 | - | 4 | 966 | |||
AK010332.1 | - | 4 | 966 | |||
AK167259.1 | - | 4 | 966 | |||
AY278951.1 | - | 4 | 843 | |||
AY455282.1 | - | 5 | 775 | |||
AY455285.1 | - | 5 | 791 | |||
BC132017.1 | - | 4 | 991 | 40130294 | ||
BC137873.1 | - | 4 | 991 | 8860892 | ||
BC144995.1 | - | 4 | 988 | 9053543 | ||
BC144996.1 | - | 4 | 991 | 9053544 |