Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031514.1 | NP_113702.1 | 3 | 318 | ||
GenBank mRNA | U13396.1 | - | 3 | 318 |