Search
Search

Pde6d| Gene Symbol: Pde6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008801.2 | NP_032827.1 | 2 | 260 | ||
| GenBank mRNA | AF039216.1 | - | 2 | 219 | ||
| AK141209.1 | - | 2 | 266 | |||
| AK165701.1 | - | 2 | 392 | |||
| AK170067.1 | - | 2 | 272 | |||
| BC005636.1 | - | 2 | 260 | 3964336 | ||