Search
Search

Rpl8| Gene Symbol: Rpl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012053.2 | NP_036183.1 | 6 | 822 | ||
| GenBank mRNA | AK088263.1 | - | 6 | 809 | ||
| AK131970.1 | - | 6 | 814 | |||
| AK145498.1 | - | 6 | 809 | |||
| AK146330.1 | - | 6 | 809 | |||
| AK151408.1 | - | 6 | 809 | |||
| AK152063.1 | - | 6 | 815 | |||
| AK167775.1 | - | 6 | 809 | |||
| AK168332.1 | - | 6 | 809 | |||
| AK168769.1 | - | 6 | 807 | |||
| AV214072.1 | - | 2 | 186 | |||
| BC043017.1 | - | 5 | 790 | 5684024 | ||
| U67771.1 | - | 5 | 794 | |||