Search
Search

Nudt7| Gene Symbol: Nudt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290181.1 | NP_001277110.1 | 2 | 1988 | ||
| NM_001290182.1 | NP_001277111.1 | 2 | 248 | |||
| NM_024437.4 | NP_077757.2 | 2 | 248 | |||
| NM_024446.5 | NP_077766.3 | 2 | 248 | |||
| GenBank mRNA | AA616044.1 | - | 1 | 90 | ||
| AF338424.1 | - | 2 | 208 | |||
| AK004924.1 | - | 2 | 1944 | |||
| AK011172.1 | - | 2 | 182 | |||
| BC033046.1 | - | 2 | 180 | |||
| BC069843.1 | - | 2 | 204 | 5102473 | ||
| CA460240.1 | - | 2 | 280 | |||