Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026487.3 | NP_080763.2 | 5 | 505 | ||
GenBank mRNA | AK009419.1 | - | 1 | 93 | ||
AK014967.1 | - | 5 | 522 | |||
AK030719.1 | - | 5 | 525 | |||
AK033639.1 | - | 5 | 529 | |||
AK044925.1 | - | 5 | 505 | |||
AK075685.1 | - | 5 | 550 | |||
AK150469.1 | - | 5 | 525 | |||
AK152059.1 | - | 5 | 525 | |||
AK165953.1 | - | 5 | 540 | |||
BC029085.1 | - | 5 | 505 | 3593451 | ||
BC043051.1 | - | 5 | 490 | 5708377 |