Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028264.4 | NP_082540.1 | 7 | 1136 | ||
GenBank mRNA | AK033845.1 | - | 7 | 1099 | ||
AK049213.1 | - | 7 | 1137 | |||
AX775959.1 | - | 7 | 1065 | |||
BC021435.1 | - | 7 | 1081 | 5029766 |