Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029420.4 | NP_083696.2 | 3 | 725 | ||
GenBank mRNA | AK010748.1 | - | 3 | 725 | ||
AK010828.1 | - | 3 | 690 | |||
AK014753.1 | - | 3 | 582 | |||
AK049139.1 | - | 3 | 726 | |||
AK053901.1 | - | 2 | 614 | |||
BC055909.1 | - | 3 | 676 | |||
BC145468.1 | - | 2 | 537 | |||
BC145688.1 | - | 3 | 590 | 40130991 |