Search
Search

| Gene Symbol: Pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172134.2 | NP_742146.1 | 1, 2 | 196 | ||
| GenBank mRNA | AK039194.1 | - | 1, 2 | 178 | ||
| AK080846.1 | - | 1, 2 | 172 | |||
| AK145470.1 | - | 1, 2 | 160 | |||
| AK166078.1 | - | 1, 2 | 179 | |||
| AK166464.1 | - | 1, 2 | 161 | |||
| BC027745.1 | - | 1, 2 | 233 | 5064695 | ||
| BC032171.1 | - | 1, 2 | 167 | |||
| Gene Symbol: Pdxk-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138248.1 | - | 1 | 1176 | ||