Search
Search

KNG1| Gene Symbol: KNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000893.3 | NP_000884.1 | 1 | 298 | ||
| NM_001102416.2 | NP_001095886.1 | 1 | 298 | |||
| NM_001166451.1 | NP_001159923.1 | 1 | 298 | |||
| GenBank mRNA | AI133186.1 | - | 1 | 208 | ||
| AK223589.1 | - | 1 | 247 | |||
| AK290839.1 | - | 1 | 197 | |||
| AK298440.1 | - | 2 | 212 | |||
| AK303768.1 | - | 1 | 278 | |||
| AK315230.1 | - | 1 | 250 | |||
| BC026253.1 | - | 1 | 67 | |||
| BC060039.1 | - | 1 | 201 | 4593077 | ||
| BG430928.1 | - | 1 | 277 | |||
| CD014130.1 | - | 1 | 260 | |||
| DA537547.1 | - | 1 | 276 | |||
| DC362362.1 | - | 1 | 298 | |||
| K02566.1 | - | 1 | 113 | |||