Search
Search

NOTCH1| Gene Symbol: NOTCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017617.4 | NP_060087.3 | 28 | 5235 | ||
| XM_011518717.2 | XP_011517019.2 | 25 | 4527 | |||
| GenBank mRNA | AB209873.1 | - | 20 | 3772 | ||
| AF308602.1 | - | 28 | 5238 | |||
| M73980.1 | - | 28 | 5238 | |||