Search
Search

| Gene Symbol: TP53BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141979.1 | NP_001135451.1 | 28 | 5918 | ||
| NM_001141980.1 | NP_001135452.1 | 28 | 5924 | |||
| NM_005657.2 | NP_005648.1 | 28 | 5944 | |||
| XM_011521985.1 | XP_011520287.1 | 27 | 5773 | |||
| XM_017022533.1 | XP_016878022.1 | 29 | 6028 | |||
| GenBank mRNA | AB210025.1 | - | 28 | 5889 | ||
| AF078776.1 | - | 28 | 5969 | |||
| AK123418.1 | - | 7 | 1930 | |||
| BC112161.1 | - | 28 | 5904 | 8327629 | ||
| BX537418.1 | - | 28 | 5923 | |||
| U09477.1 | - | 15 | 2963 | |||
| Gene Symbol: TUBGCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521454.2 | XP_011519756.1 | 17 | 4471 | ||
| XM_011521455.2 | XP_011519757.1 | 18 | 4486 | |||
| XM_017022078.1 | XP_016877567.1 | 17 | 4468 | |||