Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324095.1 | NP_001311024.1 | 4 | 523 | ||
NM_006438.4 | NP_006429.2 | 2 | 292 | |||
XM_005250756.3 | XP_005250813.1 | 2 | 193 | |||
GenBank mRNA | AB002631.1 | - | 2 | 200 | ||
AK291791.1 | - | 2 | 292 | |||
AY359038.1 | - | 2 | 216 | |||
BC103815.1 | - | 2 | 236 | 40000432 | ||
BC103816.1 | - | 2 | 236 | 40000433 |