Search
Search

PLA2G16| Gene Symbol: PLA2G16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128203.1 | NP_001121675.1 | 5 | 550 | ||
| NM_007069.3 | NP_009000.2 | 4 | 834 | |||
| XM_006718426.1 | XP_006718489.1 | 5 | 752 | |||
| XM_011544741.1 | XP_011543043.1 | 4 | 736 | |||
| GenBank mRNA | AB030814.1 | - | 4 | 460 | ||
| AB439591.1 | - | 3 | 411 | |||
| AF317086.1 | - | 4 | 465 | |||
| AI590735.1 | - | 1 | 248 | |||
| AK313075.1 | - | 4 | 833 | |||
| AX775893.1 | - | 4 | 465 | |||
| BC001387.2 | - | 4 | 766 | 3051149 | ||
| BC103807.2 | - | 4 | 555 | 40000132 | ||
| BC103808.2 | - | 4 | 505 | |||
| BG719903.1 | - | 5 | 545 | |||
| CK003948.1 | - | 5 | 551 | |||
| X92814.1 | - | 4 | 818 | |||