Search
Search

AK5| Gene Symbol: AK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012093.3 | NP_036225.2 | 11 | 2223 | ||
| NM_174858.2 | NP_777283.1 | 11 | 1612 | |||
| XM_005270739.4 | XP_005270796.1 | 11 | 1459 | |||
| XM_006710572.3 | XP_006710635.1 | 11 | 1390 | |||
| XM_017001008.1 | XP_016856497.1 | 11 | 1407 | |||
| XM_017001009.1 | XP_016856498.1 | 11 | 2115 | |||
| XM_017001010.1 | XP_016856499.1 | 11 | 2442 | |||
| XM_017001011.1 | XP_016856500.1 | 11 | 1225 | |||
| GenBank mRNA | AF062595.1 | - | 3 | 230 | ||
| AF445193.1 | - | 11 | 1612 | |||
| AY171600.2 | - | 11 | 2228 | |||
| BC012467.2 | - | 6 | 575 | |||
| BC033896.1 | - | 11 | 2228 | 4818069 | ||
| BC036666.1 | - | 11 | 1538 | 4816351 | ||
| CR541890.1 | - | 2 | 180 | |||