Search
Search

TMOD2| Gene Symbol: TMOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142885.1 | NP_001136357.1 | 8, 9 | 1140 | ||
| NM_014548.3 | NP_055363.1 | 9, 10 | 1248 | |||
| GenBank mRNA | AF177169.1 | - | 9, 10 | 1183 | ||
| AK292733.1 | - | 9, 10 | 1127 | |||
| AK293823.1 | - | 8, 9 | 1028 | |||
| BC036184.1 | - | 10, 11 | 1294 | 4815047 | ||
| BC064961.1 | - | 9, 10 | 1196 | 6141541 | ||