Search
Search

FN3KRP| Gene Symbol: FN3KRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024619.3 | NP_078895.2 | 6 | 911 | ||
| NR_046408.1 | 7 | 1039 | ||||
| GenBank mRNA | AK022233.1 | - | 6 | 878 | ||
| AK293523.1 | - | 5 | 814 | |||
| AK301958.1 | - | 7 | 1023 | |||
| AK315373.1 | - | 6 | 895 | |||
| AL136631.1 | - | 6 | 882 | |||
| AY360465.1 | - | 6 | 878 | |||
| BC001458.2 | - | 3 | 449 | |||
| BC007611.1 | - | 6 | 887 | 3351601 | ||
| BC014408.1 | - | 6 | 865 | 4652087 | ||