Search
Search

STEAP4| Gene Symbol: STEAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205315.1 | NP_001192244.1 | 4 | 955 | ||
| NM_024636.3 | NP_078912.2 | 3 | 856 | |||
| GenBank mRNA | AF423422.1 | - | 4 | 838 | ||
| AF423423.1 | - | 4 | 838 | |||
| AK026806.1 | - | 3 | 835 | |||
| AK307644.1 | - | 3 | 834 | |||