Search
Search

PSMD4| Gene Symbol: PSMD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002810.2 | NP_002801.1 | 8, 9 | 961 | ||
| XM_005245354.2 | XP_005245411.1 | 8, 9 | 991 | |||
| XM_006711464.3 | XP_006711527.1 | 7, 8 | 816 | |||
| XM_017001856.1 | XP_016857345.1 | 7, 8 | 802 | |||
| GenBank mRNA | AB033605.1 | - | 7, 8 | 1108 | ||
| BC002365.2 | - | 8, 9 | 911 | 2820813 | ||
| BC072008.1 | - | 8, 9 | 925 | 6285035 | ||
| U24704.1 | - | 8, 9 | 961 | |||
| U72664.1 | - | 6, 7 | 693 | |||