Search
Search

RAB27B| Gene Symbol: RAB27B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004163.4 | NP_004154.2 | 6 | 835 | ||
| XM_005266733.1 | XP_005266790.1 | 7 | 756 | |||
| XM_017025913.1 | XP_016881402.1 | 9 | 1589 | |||
| XM_017025914.1 | XP_016881403.1 | 8 | 984 | |||
| GenBank mRNA | AF329499.1 | - | 6 | 835 | ||
| AF498954.1 | - | 5 | 592 | |||
| AK314115.1 | - | 6 | 1113 | |||
| BC027474.1 | - | 6 | 703 | 4520182 | ||
| U57093.2 | - | 6 | 684 | |||