Search
Search

PDZRN3| Gene Symbol: PDZRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303139.1 | NP_001290068.1 | 8 | 1133 | ||
| NM_001303140.1 | NP_001290069.1 | 8 | 996 | |||
| NM_001303141.1 | NP_001290070.1 | 8 | 946 | |||
| NM_001303142.1 | NP_001290071.1 | 8 | 1126 | |||
| NM_015009.2 | NP_055824.1 | 10 | 1782 | |||
| XM_017005942.1 | XP_016861431.1 | 9 | 1777 | |||
| GenBank mRNA | AB029018.1 | - | 10 | 1782 | ||
| AK074573.1 | - | 6 | 476 | |||
| AK294761.1 | - | 8 | 937 | |||
| AK299275.1 | - | 8 | 946 | |||
| AK299560.1 | - | 8 | 980 | |||
| AK307818.1 | - | 8 | 1079 | |||
| AK316244.1 | - | 8 | 946 | |||
| AK316275.1 | - | 8 | 965 | |||
| BC150623.1 | - | 10 | 1769 | 9056993 | ||
| BC152417.1 | - | 10 | 1805 | 8862525 | ||