Search
Search

HYPM| Gene Symbol: HYPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012274.1 | NP_036406.1 | 1 | 323 | ||
| GenBank mRNA | AF049615.1 | - | 1 | 323 | ||
| AW291697.1 | - | 1 | 191 | |||
| BC104428.1 | - | 1 | 300 | 40021552 | ||
| BC104429.1 | - | 1 | 300 | 40021554 | ||
| BC113024.1 | - | 1 | 300 | 40021553 | ||
| BC113025.1 | - | 1 | 300 | 40021555 | ||