Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SETD5 SIRNA Assays ›
          Gene Symbol:
          SETD5
          Gene Name:
          SET domain containing 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 9397700 - 9478154 on Build GRCh38
          UniGene ID:
          Hs.288164
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS147922
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SETD5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001080517.2 NP_001073986.1 18 3094
          NM_001292043.1 NP_001278972.1 20 3358
          XM_005265301.1 XP_005265358.1 18 3132
          XM_005265303.1 XP_005265360.1 20 3513
          XM_011533920.1 XP_011532222.1 21 3608
          XM_011533921.1 XP_011532223.1 20 3210
          XM_011533922.1 XP_011532224.1 19 3171
          XM_011533927.1 XP_011532229.1 21 3551
          XM_011533928.1 XP_011532230.1 19 3114
          XM_011533930.1 XP_011532232.1 21 3552
          XM_011533932.1 XP_011532234.1 19 3319
          XM_011533933.1 XP_011532235.1 17 2688
          XM_017006767.1 XP_016862256.1 21 5270
          XM_017006768.1 XP_016862257.1 19 4784
          XM_017006769.1 XP_016862258.1 19 6887
          XM_017006770.1 XP_016862259.1 20 6926
          XM_017006771.1 XP_016862260.1 18 3132
          XM_017006772.1 XP_016862261.1 19 6887
          XM_017006773.1 XP_016862262.1 19 6830
          XM_017006774.1 XP_016862263.1 18 3075
          XM_017006775.1 XP_016862264.1 19 6830
          XM_017006776.1 XP_016862265.1 20 3415
          XM_017006777.1 XP_016862266.1 20 3358
          XM_017006778.1 XP_016862267.1 21 3399
          XM_017006779.1 XP_016862268.1 19 3376
          XM_017006780.1 XP_016862269.1 20 3360
          XM_017006781.1 XP_016862270.1 21 3399
          XM_017006782.1 XP_016862271.1 20 6926
          XM_017006783.1 XP_016862272.1 22 3624
          XM_017006784.1 XP_016862273.1 20 6926
          GenBank mRNA AB051544.1 - 14 2888
          AK001569.1 - 7 1277
          AK024384.1 - 6 1188
          AK025478.1 - 2 202
          AK091393.1 - 15 2397
          AK092046.1 - 20 3313
          AK127298.1 - 8 1981
          AK289529.1 - 9 1676
          AL442073.1 - 5 1074
          BC020956.1 - 7 1308 3851178
          BX648380.1 - 10 1696
          CR627408.1 - 17 2710

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106614, XM_006237014, XM_006237012, XM_008763189, XM_006237018, and others.
          This siRNA also targets the following transcripts in Mouse: NM_028385, AK053541, BC083184, AK122551, AK049143, and others.

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0