Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD19P SIRNA Assays ›
          Gene Symbol:
          ANKRD19P EEF1D
          Gene Name:
          ankyrin repeat domain 19, pseudogene
          eukaryotic translation elongation factor 1 delta
          Gene Aliases:
          ANKRD19, bA526D8.2
          EF-1D, EF1D, FP1047
          Chromosome Location:
          Chr.9: 92809611 - 92838457 on Build GRCh38
          Chromosome Location:
          Chr.8: 143579697 - 143597675 on Build GRCh38
          UniGene ID:
          Hs.643597
          Hs.333388
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS151386
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD19P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026868.1 9 1868
          GenBank mRNA AK093497.1 - 7 1460
          BC028712.1 - 9 1832
          Gene Symbol: EEF1D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130053.2 NP_001123525.2 5 1603
          NM_001130055.2 NP_001123527.1 4 674
          NM_001130057.2 NP_001123529.1 3 680
          NM_001195203.1 NP_001182132.1 3 498
          NM_001289950.1 NP_001276879.1 3 495
          NM_001960.4 NP_001951.2 3 498
          NM_032378.4 NP_115754.3 5 1720
          XM_005250820.3 XP_005250877.1 4 6595
          XM_005250823.1 XP_005250880.1 4 1329
          XM_006716519.3 XP_006716582.1 4 6592
          XM_006716522.1 XP_006716585.1 4 1583
          XM_006716523.1 XP_006716586.1 5 1543
          XM_006716524.1 XP_006716587.1 4 1350
          XM_006716525.1 XP_006716588.1 3 518
          XM_011516905.2 XP_011515207.1 5 6165
          XM_011516906.2 XP_011515208.1 7 4757
          XM_017013168.1 XP_016868657.1 5 5596
          XM_017013169.1 XP_016868658.1 6 5252
          XM_017013170.1 XP_016868659.1 5 5038
          XM_017013171.1 XP_016868660.1 4 1326
          GenBank mRNA AF370363.1 - 5 1303
          AK024550.1 - 5 1457
          AK093756.1 - 3 1640
          AK225040.1 - 5 1458
          AK225243.1 - 3 235
          AK225270.1 - 3 236
          AK293339.1 - 3 236
          AK314311.1 - 5 1339
          BC007847.2 - 5 1324 4129368
          BC009907.2 - 1 220
          BC012819.2 - 1 220
          BC062535.1 - 3 212
          BC071840.1 - 3 200
          BG748894.1 - 3 680
          BI667117.1 - 3 503
          BQ064320.1 - 355
          BQ686171.1 - 4 393
          BT007242.1 - 3 1261
          CF128478.1 - 3 585
          CN424083.1 - 3 439
          CN424105.1 - 3 283
          CX787126.1 - 3 545
          Z21507.1 - 3 232

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          translation elongation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          translational elongation
          signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          cellular response to ionizing radiation
          cell-cell adhesion
          DNA binding
          translation elongation factor activity
          signal transducer activity
          protein binding
          translation factor activity, RNA binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0