Search
Search

FAM101A
ZNF664-FAM101A| Gene Symbol: FAM101A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181709.4 | NP_859060.3 | 3 | 419 | ||
| GenBank mRNA | AK074545.1 | - | 5 | 452 | ||
| AK126577.1 | - | 3 | 419 | |||
| BC042102.1 | - | 3 | 326 | |||
| BC141805.1 | - | 2 | 187 | 40147249 | ||
| BQ182211.1 | - | 1 | 482 | |||
| Gene Symbol: ZNF664-FAM101A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204299.1 | NP_001191228.1 | 5 | 682 | ||