Search
Search

LYPD4| Gene Symbol: LYPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173506.6 | NP_775777.3 | 3 | 1351 | ||
| XM_005258543.3 | XP_005258600.1 | 3 | 379 | |||
| XM_005278383.4 | XP_005278440.1 | 3 | 1390 | |||
| XM_005278385.4 | XP_005278442.1 | 3 | 1390 | |||
| XM_005278387.4 | XP_005278444.1 | 3 | 1390 | |||
| XM_005278388.3 | XP_005278445.1 | 2 | 1174 | |||
| XM_006723035.2 | XP_006723098.1 | 2 | 167 | |||
| XM_011526483.2 | XP_011524785.1 | 3 | 547 | |||
| XM_011526484.2 | XP_011524786.1 | 3 | 452 | |||
| GenBank mRNA | AK292305.1 | - | 3 | 1346 | ||
| AY358726.1 | - | 3 | 1120 | |||
| AY771680.1 | - | 2 | 137 | |||