Search
Search

EXT2| Gene Symbol: EXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000401.3 | NP_000392.3 | 6 | 1186 | ||
| NM_001178083.1 | NP_001171554.1 | 6 | 1365 | |||
| NM_207122.1 | NP_997005.1 | 6 | 1365 | |||
| XM_011519950.1 | XP_011518252.1 | 7 | 1184 | |||
| XM_011519951.1 | XP_011518253.1 | 7 | 1294 | |||
| GenBank mRNA | AK296713.1 | - | 6 | 879 | ||
| AK312375.1 | - | 6 | 1242 | |||
| BC010058.1 | - | 6 | 1239 | 3947503 | ||
| BC068545.1 | - | 6 | 1205 | 5263866 | ||
| BX648142.1 | - | 6 | 1163 | |||
| U62740.1 | - | 6 | 1365 | |||
| U64511.1 | - | 5 | 1057 | |||
| U72263.1 | - | 6 | 1197 | |||