Search
Search

MAPRE1| Gene Symbol: MAPRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012325.2 | NP_036457.1 | 2 | 217 | ||
| XM_011528696.2 | XP_011526998.1 | 2 | 219 | |||
| GenBank mRNA | AK312590.1 | - | 2 | 134 | ||
| BC040410.1 | - | 2 | 144 | |||
| BC106068.1 | - | 2 | 147 | 6527006 | ||
| BC109281.3 | - | 2 | 141 | 40031641 | ||
| BC128442.1 | - | 2 | 112 | |||
| BJ992159.1 | - | 2 | 217 | |||
| U24166.1 | - | 2 | 142 | |||