Search
Search

TBC1D15| Gene Symbol: TBC1D15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146213.1 | NP_001139685.2 | 7 | 914 | ||
| NM_001146214.1 | NP_001139686.1 | 8 | 1006 | |||
| NM_022771.4 | NP_073608.4 | 8 | 965 | |||
| NR_027449.1 | 6 | 703 | ||||
| XM_006719564.2 | XP_006719627.1 | 8 | 947 | |||
| XM_006719565.3 | XP_006719628.1 | 8 | 885 | |||
| XM_006719566.2 | XP_006719629.1 | 9 | 1096 | |||
| XM_006719567.2 | XP_006719630.1 | 7 | 782 | |||
| XM_011538679.1 | XP_011536981.1 | 10 | 1022 | |||
| XM_011538680.1 | XP_011536982.1 | 8 | 980 | |||
| XM_011538681.2 | XP_011536983.1 | 8 | 929 | |||
| XM_011538682.1 | XP_011536984.1 | 7 | 784 | |||
| XM_017019824.1 | XP_016875313.1 | 9 | 982 | |||
| XM_017019825.1 | XP_016875314.1 | 8 | 1001 | |||
| XM_017019826.1 | XP_016875315.1 | 8 | 931 | |||
| XM_017019827.1 | XP_016875316.1 | 5 | 605 | |||
| XM_017019828.1 | XP_016875317.1 | 6 | 723 | |||
| GenBank mRNA | AB449901.1 | - | 7 | 839 | ||
| AK022147.1 | - | 7 | 862 | |||
| AK225221.1 | - | 7 | 843 | |||
| AK225747.1 | - | 7 | 860 | |||
| AK292306.1 | - | 6 | 634 | |||
| AK307922.1 | - | 8 | 931 | |||
| AL833639.1 | - | 6 | 642 | |||
| BC028352.2 | - | 8 | 913 | 4827246 | ||