Search
Search

Rcc2| Gene Symbol: Rcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173867.5 | NP_776292.1 | 8 | 1121 | ||
| GenBank mRNA | AK076040.1 | - | 8 | 1052 | ||
| AK076046.1 | - | 1352 | ||||
| AK129370.1 | - | 7 | 825 | |||
| BC020385.1 | - | 6 | 654 | |||
| BC034759.1 | - | 5 | 440 | |||
| BC060624.1 | - | 8 | 1075 | |||
| BC086666.1 | - | 8 | 1044 | 30634817 | ||