Search
Search

Idh1| Gene Symbol: Idh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111320.1 | NP_001104790.1 | 8 | 1223 | ||
| NM_010497.3 | NP_034627.3 | 8 | 1201 | |||
| GenBank mRNA | AF020039.1 | - | 7 | 1051 | ||
| AK087063.1 | - | 8 | 1103 | |||
| AK149019.1 | - | 8 | 1132 | |||
| AK151212.1 | - | 8 | 1126 | |||
| AK159173.1 | - | 8 | 1132 | |||
| AK160896.1 | - | 8 | 1133 | |||
| AK167158.1 | - | 8 | 1173 | |||
| AK167583.1 | - | 8 | 1137 | |||
| AK168832.1 | - | 8 | 1174 | |||
| AK216686.1 | - | 2 | 57 | |||
| BC088986.1 | - | 8 | 1173 | 6808731 | ||
| DQ403075.1 | - | 7 | 951 | |||