Search
Search

Kif9| Gene Symbol: Kif9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163569.1 | NP_001157041.1 | 16 | 1774 | ||
| NM_010628.3 | NP_034758.2 | 16 | 1779 | |||
| GenBank mRNA | AJ132889.1 | - | 16 | 1712 | ||
| AK029506.1 | - | 16 | 1672 | |||
| AK029776.1 | - | 16 | 1744 | |||
| AK077169.1 | - | 8 | 715 | |||
| AK087008.1 | - | 16 | 1744 | |||
| BC115765.1 | - | 17 | 1972 | 40059805 | ||