Search
Search

Nedd4| Gene Symbol: Nedd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010890.3 | NP_035020.2 | 19 | 1946 | ||
| GenBank mRNA | AK088620.1 | - | 19 | 1850 | ||
| AK088767.1 | - | 19 | 1859 | |||
| AK122203.1 | - | 19 | 1789 | |||
| AK133838.1 | - | 19 | 1859 | |||
| BC138813.1 | - | 19 | 1824 | 8861835 | ||
| D85414.1 | - | 19 | 1946 | |||
| U96635.1 | - | 19 | 2017 | |||