Search
Search

Polh| Gene Symbol: Polh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030715.3 | NP_109640.1 | 2 | 342 | ||
| GenBank mRNA | AB027128.1 | - | 2 | 368 | ||
| AK036296.1 | - | 2 | 343 | |||
| BC049159.1 | - | 2 | 313 | |||
| BC128366.1 | - | 2 | 339 | 40111620 | ||