Search
Search

C2cd4c| Gene Symbol: C2cd4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168624.1 | NP_001162095.1 | 2 | 1272 | ||
| NM_198614.3 | NP_941016.2 | 2 | 1398 | |||
| GenBank mRNA | AK029946.1 | - | 2 | 1273 | ||
| BC089008.1 | - | 2 | 1380 | 30537363 | ||