Search
Search

Oas1b| Gene Symbol: Oas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083925.1 | NP_001077394.1 | 4 | 783 | ||
| GenBank mRNA | AB067529.1 | - | 4 | 783 | ||
| AF328926.1 | - | 4 | 783 | |||
| AF418004.1 | - | 4 | 783 | |||
| AF418005.1 | - | 4 | 783 | |||
| AF418006.1 | - | 4 | 783 | |||
| AF466822.1 | - | 4 | 740 | |||
| AF466823.1 | - | 4 | 740 | |||
| AK080152.1 | - | 4 | 776 | |||
| BC012877.1 | - | 4 | 775 | 4016177 | ||
| X55982.1 | - | 3 | 546 | |||