Search
Search

Pvalb| Gene Symbol: Pvalb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013645.3 | NP_038673.2 | 4 | 340 | ||
| GenBank mRNA | AK002354.1 | - | 4 | 348 | ||
| AK005371.1 | - | 4 | 338 | |||
| AK013561.1 | - | 4 | 332 | |||
| AK131986.1 | - | 4 | 410 | |||
| BC027424.1 | - | 4 | 340 | 4925213 | ||
| X67141.1 | - | 1 | 15 | |||