Search
Search

Dlg3| Gene Symbol: Dlg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177778.2 | NP_001171249.1 | 18 | 2601 | ||
| NM_001177779.1 | NP_001171250.1 | 12 | 1598 | |||
| NM_001177780.1 | NP_001171251.1 | 10 | 990 | |||
| NM_001290402.1 | NP_001277331.1 | 17 | 2547 | |||
| NM_016747.4 | NP_058027.1 | 19 | 2643 | |||
| GenBank mRNA | AK122474.1 | - | 18 | 2600 | ||
| AK145815.1 | - | 12 | 1353 | |||
| AK148464.1 | - | 10 | 970 | |||
| AK159655.1 | - | 10 | 978 | |||
| AK167365.1 | - | 10 | 977 | |||
| BC060713.1 | - | 17 | 2675 | |||
| BC094368.1 | - | 17 | 2542 | 6842105 | ||
| D87117.1 | - | 19 | 2617 | |||