Search
Search

Hecw1| Gene Symbol: Hecw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081348.3 | NP_001074817.3 | 3, 4 | 789 | ||
| GenBank mRNA | AB083710.1 | - | 1, 2 | 292 | ||
| AK047678.1 | - | 3, 4 | 724 | |||
| AK051569.1 | - | 2, 3 | 610 | |||
| AK053694.1 | - | 2, 3 | 734 | |||
| AK084246.1 | - | 2, 3 | 610 | |||