Search
Search

Chi3l1| Gene Symbol: Chi3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053560.1 | NP_446012.1 | 8 | 815 | ||
| XM_006249892.2 | XP_006249954.2 | 8 | 869 | |||
| XM_008769574.1 | XP_008767796.1 | 8 | 836 | |||
| GenBank mRNA | AF062038.1 | - | 6 | 696 | ||
| BC091365.1 | - | 8 | 815 | 7319171 | ||
| EF467168.1 | - | 6 | 696 | |||
| FQ232458.1 | - | 5 | 481 | |||