Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033975.1 | NP_001029147.1 | 1 | 155 | ||
NM_012858.2 | NP_036990.1 | 2 | 116 | |||
XM_006228975.2 | XP_006229037.1 | 5 | 1921 | |||
GenBank mRNA | CB566905.1 | - | 119 | |||
CB812721.1 | - | 1 | 156 | |||
D00576.1 | - | 1 | 98 | |||
U25653.1 | - | 5 | 1926 | |||
U25802.1 | - | 5 | 1927 | |||
V01542.1 | - | 2 | 108 |