Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105966.1 | NP_001099436.1 | 2 | 431 | ||
XM_008769625.1 | XP_008767847.1 | 2 | 555 | |||
GenBank mRNA | BC166446.1 | - | 2 | 445 | 8364240 | |
FQ226272.1 | - | 2 | 417 | |||
FQ227495.1 | - | 2 | 435 | |||
FQ230214.1 | - | 2 | 464 | |||
FQ230343.1 | - | 2 | 429 | |||
FQ231443.1 | - | 2 | 418 | |||
FQ232342.1 | - | 2 | 435 | |||
FQ234634.1 | - | 2 | 464 | |||
FQ234923.1 | - | 2 | 428 |